Learn More
Gibco™ Floor Plate Cell Expansion Kit
Product Code. 15631215 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 pack
Product Code. Quantity Unit Size
15631215 1 kit 1 pack
1 options
This item is not returnable. View return policy
Product Code. 15631215 Supplier Gibco™ Supplier No. A3165801
 Restricted Item
Fecha estimada de envoi: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Floor Plate Cell Expansion Kit is a component of the Gibco™ PSC Dopaminergic Neuron Differentiation Kit that is also offered separately.
It enables additional expansion of floor plate progenitor cells generated using complete Floor Plate Cell Specification Medium (part of the Differentiation kit).
Note: Each additional expansion beyond passage 2 will result in an additional 4–6-fold increase in the total number of floor plate cells without compromising the quality of TH neurons (positive for both TH and FOXA2). Further expansion (up to passage 10) results in stable expansion of progenitor cells, but the resulting TH neurons may show decreased FOXA2 expression.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Content And Storage • 1 x 500 mL Floor Plate Cell Expansion Base
• 1 x 10 mL Floor Plate Cell Expansion Supplement
Cell Type Stem Cells (Neural)
Shipping Condition Dry ice and gel pack
For Research Use Only. Not for use in diagnostic procedures.