Learn More
Scotch-Brite™ Floor Pads, Red
Product Code. 17198840
Change view
Click to view available options
Diameter (Metric):
203 mm
254 mm
280 mm
305 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
584 mm
Unit Size:
Pack of 10
Pack of 5
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
17198840 406 mm Pack of 5
17168850 203 mm Pack of 10
17138840 254 mm Pack of 5
17128850 480 mm Pack of 5
17138850 505 mm Pack of 5
17148850 530 mm Pack of 5
17158850 584 mm Pack of 5
17148840 280 mm Pack of 5
17158840 305 mm Pack of 5
17168840 330 mm Pack of 5
17178840 355 mm Pack of 5
17188840 380 mm Pack of 5
17108850 432 mm Pack of 5
17118850 460 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 17198840 Supplier Scotch-Brite™ Supplier No. 7100182728
  • ONLINE EXCLUSIVE PRICE
    122.00€
    74.59€ / paquete de 5
    Ahorro 47.41€
    38% Descuento
Fecha estimada de envoi: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Designed for everyday cleaning and light scrubbing to remove light soil, scuff marks, and black heel marks. These pads clean when damp and buff when dry. Open-web construction means this washable, reusable pad resists clogging and loading.
  • For everyday cleaning and light scrubbing
  • Removes light soil, scuff marks, and black heel marks
  • Cleans when damp and buffs when dry
  • Conformable fibers provide maximum pad-to-floor contact
  • Reversible, washable pad resists clogging and loading
  • Consistent, proven performance with long floor pad life
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Pad
Color Red
Diameter (Metric) 406 mm
For Use With (Application) Everyday Cleaning, Light Scrubbing