missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLI1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11347-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
ETS-1 is the prototype member of a family of genes identified on the basis of homology to the v-Ets oncogene isolated from the E26 erythroblastosis virus. This family of genes currently includes ETS-1, ETS-2, ERG-1, ERG-2, ELK, E74, FLI-1, PU. 1 and PEA3. Members of the ETS gene family exhibit varied pattern of tissue expression, and share a highly conserved carboxy terminal domain containing a sequence related to the SV40 large T antigen nuclear localization signal sequence.Especificaciones
FLI1 | |
Polyclonal | |
Unconjugated | |
FLI1 | |
EWSR2, FLI1, Proto oncogene Fli 1, SIC 1, Transcription factor ERGB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14247, 2313 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P26323, Q01543 | |
FLI1 | |
FLI1 Fusion Protein Ag1899 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |