Learn More
Fleroxacin, TRC
Product Code. 30398529
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30398529 50 mg 50mg
1 options
This item is not returnable. View return policy
Product Code. 30398529 Supplier Toronto Research Chemicals Supplier No. TRCF40125050MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fleroxacin
CAS 79660-72-3
Molecular Formula C17 H18 F3 N3 O3
Quantity 50 mg
Synonym 3-Quinolinecarboxylic acid, 6,8-difluoro-1-(2-fluoroethyl)-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-, 6,8-Difluoro-1-(2-fluoroethyl)-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, AM 833, Flerofloxacin, Fleroxacin, Megalocin, Megalone, Quinodis, Ro 23-6240, Ro 23-6240/000
InChI Formula InChI=1S/C17H18F3N3O3/c1-21-4-6-22(7-5-21)15-12(19)8-10-14(13(15)20)23(3-2-18)9-11(16(10)24)17(25)26/h8-9H,2-7H2,1H3,(H,25,26)
SMILES CN1CCN(CC1)c2c(F)cc3C(=O)C(=CN(CCF)c3c2F)C(=O)O
IUPAC Name 6,8-difluoro-1-(2-fluoroethyl)-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
Molecular Weight (g/mol) 369.34
Formula Weight 369.13