Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Amino
Product Code. 16350176 Shop All Buchi Products
Change view
Click to view available options
Size (Metric) Tube Rack:
12 g
Includes:
5 Pc.
Product Code. Size (Metric) Tube Rack Includes
16350176 12 g 5 Pc.
1 options
Product Code. 16350176 Supplier BUCHI Supplier No. 140000060
 Restricted Item
Fecha estimada de envoi: 16-12-2026
Add to basket
Add to basket
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
  • Most economical solution without sacrificing quality for standard separations
  • Large selection of cartridge sizes from 4g-5,000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
  • Silica is ideal for basic applications and polar compounds using organic solvents
  • Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
  • C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
  • Neutral Alumina is ideal for aldehydes, ketones, and esters
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Type Amino
Includes 5 Pc.
Resolution 40 to 60 μm
Size (Metric) Tube Rack 12 g
Column Type Amino
For Use With (Application) Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates
For Use With (Equipment) Any Flash System