Learn More
Fisherbrand™ Black Phenolic PolyCone Cap
Forms an exceptionally tight seal and offer a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.
Marca:  Fisherbrand™
Detalles adicionales : Peso : 1.50000kg
 Ver más versiones de este producto
Código de producto. 15553261
  • 258.00€ / paquete de 576
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Liner molds itself around the sealing areas of finish as cap is screwed on
  • Eliminates leakage, evaporation, contamination, binding and backing off
Especificación
Especificación
Phenolic
Phenolic
Cap, PE Cone Lined
22/400
22-400
Polyseal
576/Cs.
Black
Polycone
screw cap
Videos
Documentación