Learn More
Fisherbrand™ Black Phenolic PolyCone Cap
Forms an exceptionally tight seal and offers a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.
Marca:  Fisherbrand™
Detalles adicionales : Peso : 1.50000kg
 Ver más versiones de este producto
Código de producto. 15563261
  • 257.00€ / paquete de 576
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Liner molds itself around sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off
  • Great for everything from liquids, to soaps, to macro-invertebrates
Especificación
Especificación
ISO 9001
Polyseal
Phenolic, Phenolic
Packed in 48 bags of 12
Cap, PE Cone Lined, PolyCone Lined Cap
Black
18-400
Screw
Phenolic
Black, Black
48 Cs.
576EA/Cs.
Bottle
PolyCone
Round
Videos
Documentación