Learn More
Fisherbrand™ Histosette™ II Tissue Processing/Embedding Cassettes with Separate Lid and Base
Fisherbrand™ Histosette™ II Tissue Processing/Embedding Cassettes with Separate Lid and Base open and close as often as necessary — cassettes will always relock securely.
Marca:  Fisherbrand™
Detalles adicionales : Peso : 0.80000kg
 Ver más versiones de este producto
Código de producto. 10788191
  • 250.00€ / paquete de 1500
Fecha estimada de envío
23-07-2020
Añadir a la cesta
Descripción
Descripción
  • Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
  • Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Especificaciones
Especificaciones
Slots
Combination Lid
Plastic
45°
Tan
Biopsy Cassette