Learn More
Fisherbrand™ Graduated Valumetric™ Bottles without Caps
Catalog Number 11543752
Click to view available options
Capacidad (métrico):
60 mL
125 mL
Store a variety of liquids and solids—dry powders, specimens and soil samples
  • Clear, USP Type III soda-lime glass
  • Highly visible, white screened graduations in millimeters and ounces for general mixing and measuring applications
  • Wide mouth for easy entry and retrieval during storage and packaging applications
Capacidad 2 oz., 60mL
Tamaño del tapón 38-400 mm
Tipo de tapón Screw Cap
Capacidad (métrico) 60 mL
Capacidad (imperial) 2 oz.
Color Clear
Material Soda Lime Glass
Cantidad 288/PK.
Closure Included No
Boca Wide