Learn More
Fisherbrand™ Combs for Midi-Plus Wide Format Horizontal Gel Units, 2mm Thick
For use with Midi-Plus Wide Format Horizontal Gel Units.
Marca:  Fisherbrand
Detalles adicionales : Peso : 0.02500kg
 Ver más versiones de este producto
Código de producto. 11523412
  • 43.30€ / cada uno
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximises sample volume
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 12-sample, 25-sample and 50-sample combs are Multichannel pipettor compatible
Especificación
Especificación
2mm
Comb
1 Each
30μL
Blue
Midi-Plus Wide Format Horizontal Gel Units
50
Videos