Learn More
Fisher BioReagents™ SurePrep™ Urine Bacterial RNA Purification Kit
Description
Includes
Lysis solution (40mL); Wash solution (22mL); RNA elution buffer (6mL); Spin columns (50); Collection tubes (50); Elution tubes (1.7mL) (50); Product insert (1)
- Fast and easy processing using rapid spin-column format
- Isolate total RNA, from large mRNA down to microRNA (miRNA)
- No phenol or chloroform extractions
- Isolate RNA from both Gram negative and Gram positive bacteria, including E. coli, Proteus spp., Klebsiella spp., Enterobacter spp., Serratia spp., Pseudomonas spp., Clostridial ssp. and Leptospirosis spp., as well as Chlamydia trachomatis and Neisseria gonorrhoeae
Purified bacterial RNA suitable for real-time PCR, reverse transcription PCR, Northern blotting, RNase protection, primer extension, and expression array assays.
Specifications
Specifications
| Format | Spin Column |
| For Use With (Application) | Purified bacterial RNA suitable for RT-PCR, reverse transcription PCR, northern blotting, RNase protection, primer extension and expression array assays |
| Includes | Lysis solution 40mL, wash solution 22mL, RNA Elution Buffer 6mL, spin columns 50, collection tubes 50, 50 elution tubes 1.7mL, product insert 1 |
| Quantity | 50 Preps |
| Yield | ≈1μg RNA/1 x 105 cells |