Learn More
GVS Filter Technology SPR404 Elipse Integra Safety Goggle
Click to view available options
:
GVS SPR404 Integra Safety Goggle
Descripción
This mask is recommended for dust and fumes containing substances such as micro-organisms, marble, gypsum, titanium oxide, soapstone, rock wool, wood, detergents, textile fibers, spices, salt, animal feeds, etc..
- Protects against dust that can cause lung disease. In particular, protects against coal, silica, cotton, iron ore, graphite, kaolin, zinc, aluminium dusts.
- Protects against harmful dusts such as asbestos, bauxite, coal, silica, iron, and against toxic dusts such as manganese, lead and chromium.
- Additionally the filters remove small amounts of VOC characterised by smell/nuisance odor.
- Extremely lightweight with integrated eye and respirator protection to give a better face fit, more comfort, and reduce the risk of mist building up inside the lens.
- Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.
- Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
Especificaciones
Especificaciones
Color | Azul |
Desechables | Reusable |