Learn More
Olympus™ Filter Holder, CH2-FH
Use this to hold a wide variety of filters for different conditions. Olympus™ Filter Holder CH2-FH accepts filters for daylight, color-temperature conversion, heal absorption, color correction, sharp cut-off, light dispersion and others.
Marca:  Olympus™ K10017074
Detalles adicionales : Peso : 0.01000kg
Código de producto. 12553856
  • 18.40€ / cada uno
Fecha estimada de envío
23-07-2020
Añadir a la cesta
Descripción
Descripción
  • Accepts a 32.5mm diameter filter.
  • Accepts filters for the following:
    • Daylight.
    • Color-temperature conversion.
    • Heat absorption.
    • Color correction.
    • Sharp cut-off.
    • Light dispersion.
Especificaciones
Especificaciones
Filters
CH2 microscopes