missing translation for 'onlineSavingsMsg'
Learn More

Ficolin-2 Antibody (19), FITC, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-05042F

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213484

  • 519.42€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Ficolin-2 Monoclonal specifically detects Ficolin-2 in Human samples. It is validated for ELISA.
Especificaciones

Especificaciones

Ficolin-2
Monoclonal
FITC
PBS with 0.05% Sodium Azide
FCN2
Purified procaryot expressed recombinant human L-Ficolin and full-length L-Ficolin (Ficolin-2) protein
0.1 mL
Primary
Reacts specifically with L-Ficolin. No cross reactivity is seen with M-Ficolin (Ficolin-1) and H-Ficolin (Ficolin-3).
Store at 4C in the dark.
IgG2a κ
ELISA
19
ELISA
37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin-B, Ficolin-beta, Hucolin, L-ficolin, P35, Serum lectin p35
Mouse
Protein A purified
RUO
2220
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales