missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIBCD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25125-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
FIBCD1 is a single-pass membrane protein. It contains 1 fibrinogen C-terminal domain. The exact function of FIBCD1 remains unknown.Especificaciones
FIBCD1 | |
Polyclonal | |
Unconjugated | |
FIBCD1 | |
FIBCD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84929 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.73 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N539 | |
FIBCD1 | |
FIBCD1 Fusion Protein Ag18355 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |