Learn More
Thermo Scientific™ FHZ 632L-10 Gamma Detector
Product Code. 18867941 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Dose Rate Detector
Unit Size:
Each
Product Code. Product Type Unit Size
18867941 Dose Rate Detector Each
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 18867941 Supplier Thermo Scientific™ Supplier No. 4254057
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
The FHZ 632 L-10 Gamma Detector with a proportional counter tube is used for highly sensitive dose rate measurement of X-ray and gamma radiation.
The FHZ 632 L-10 Gamma Detector with a proportional counter tube is used for highly sensitive dose rate measurement of X-ray and gamma radiation.
Measuring Range: 10 μR/h - 10 R/h (100 nSv/h - 100 mSv/h). Sensitivity: Approx. 20 s-1/(mR/h) [2 s-1/(μSv/h)]. Energy Range 30 keV - 4.4 MeV. Energy response according to ambient dose equivalent rate H*(10).
TRUSTED_SUSTAINABILITY
Specifications
Description FHZ 632 L--10 Dose Rate Detector
Detector Type Proportional
Measurement Range 10 nSv/h to 100 mSv/h
Sensitivity 2 cps/(µSv/h)
Weight (Metric) 185 g
Energy Range 30 keV to 4.4 MeV
Range IP67
Storage Temperature (Metric) - 40°C to + 70°C
Product Type Dose Rate Detector
Dimension (Metric) Diameter: 35 mm Length: 162 mm (without plug-in connection)
Operating Limit Temperature Range (Metric) - 30°C to + 55°C
Protection Class IP67