Learn More
FGFR1 Antibody (M19B2), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NB600-1287IR
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214213
  • 589.62€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
FGFR1 Monoclonal specifically detects FGFR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Especificaciones
Especificaciones
FGFR1
Monoclonal
DyLight 755
P11362
FGFR1
Recombinant human ectodomain of FGFR1b expressed in E. coli beginning with proline 23; antigen contained NH2-terminal gly-ser-pro-gly-ile and COOH-terminal glu-phe sequences.
0.1 mL
Angiogenesis, Apoptosis, Core ESC Like Genes, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers
2260
Human, Mouse, Rat
Purified
Western Blot, Flow Cytometry
M19B2
Western Blot, Flow Cytometry
basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, FLT-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2
Mouse
Protein G purified
RUO
Primary
Reacts with both alpha and beta isoforms.
Store at 4C in the dark.
IgG1