missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF18 Mouse anti-Human, Mouse, Clone: 1D11D12, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60341-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
FGF18 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. It has been shown in vitro that this protein is able to induce neurite outgrowth in PC12 cells. Studies of the similar proteins in mouse and chick suggested that this protein is a pleiotropic growth factor that stimulates proliferation in a number of tissues, most notably the liver and small intestine. Knockout studies of the similar gene in mice implied the role of this protein in regulating proliferation and differentiation of midline cerebellar structures.Especificaciones
FGF18 | |
Monoclonal | |
0.74 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O76093, O89101 | |
FGF18 | |
FGF18 Fusion Protein Ag2041 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Western Blot | |
1D11D12 | |
Unconjugated | |
FGF18 | |
FGF 18, FGF18, fibroblast growth factor 18, ZFGF5 | |
Mouse | |
Protein A | |
RUO | |
14172, 8817 | |
-20°C | |
Liquid |