Learn More
Novus Biologicals™ FCF1 Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-10653
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18216195
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
The FCF1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC21281 and based on accession number NM_15962. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens FCF1 small subunit (SSU) processome component homolog (S. cerevisiae) (FCF1), mRNA.
Native
Certificados
Ofertas especiales
Ofertas especiales
For Research Use Only