missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FCER2 (Human) Recombinant Protein
Human FCER2 full-length ORF ( AAH14108, 1 a.a. - 321 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002208-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MEEGQYSEIEELPRRRCCRRGTQIVLLGLVTAALWAGLLTLLLLWHWDTTQGLKQLEERAARNVSQVSKNLESHHGDQMAQKSQSTQISQELEELRAEQQRLKSQDLELSWNLNGLQADLSSFKSQELNERNEASDLLERLREEVTKLRMELQVSSGFVCNTCPEKWINFQRKCYYFGKGTKQWVHARYACDDMEGQLVSIHSPEEQDFLTKHASHTGSWIGLRNLDLKGEFIWVDGSHVDYSNWAPGEPTSRSQGEDCVMMRGSGRWNDAFCDRKLGAWVCDRLATCTPPASEGSAESMGPDSRPDPDGRLPTPSAPLHS
Especificaciones
AAH14108 | |
Fc fragment of IgE, low affinity II, receptor for (CD23) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FCER2 | |
GST |
2208 | |
Wheat germ expression system | |
10 μg | |
CD23, CD23A, CLEC4J, FCE2, IGEBF | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |