missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RI/CD64 Antibody (1), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90403G

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215318

  • 605.82€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Fc gamma RI/CD64
Monoclonal
DyLight 488
50mM Sodium Borate
Rabbit
Protein A purified
RUO
2209
Mouse
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
1
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Fc gamma RI/CD64 extracellular domain (NP_034316.1; Met 1-Pro 297).
0.1 mL
Primary
Has cross-reactivity in ELISA with: Human CD64/ FCGR1A No cross-reactivity in ELISA with: Mouse CD16-2/FCGR4 Mouse FCGR2B/ CD32 Mouse FCGR3/ CD16 Human CD16a/ FCGR3A Human CD16b/ FCGR3B Human CD32a/ FCGR2A Human CD32b/ FCGR2B
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales