missing translation for 'onlineSavingsMsg'
Learn More

FAT10 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP2-81752DL594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215277

  • 577.74€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

FAT10 Polyclonal antibody specifically detects FAT10 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones

Especificaciones

FAT10
Polyclonal
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 594
Diubiquitin, FAT10diubiquitin, GABBR1, UBD-3, ubiquitin D, Ubiquitin-like protein FAT10
FAT10 antibody was raised against a 16 amino acid peptide near the center of human FAT10. The immunogen is located within amino acids 80 - 130 of FAT10. Amino Acid Squence: ESGDEAKRHLLQVRRS
0.1 mL
Cell Biology, Immunology
10537
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales