Learn More
Checkers Fastlane™ Antislip Rubber Pad Kit
Product Code. 17732508
Change view
Click to view available options
Color:
Black
Unit Size:
Each
Product Code. Color Unit Size
17732508 Black Each
1 options
This item is not returnable. View return policy
Product Code. 17732508 Supplier Checkers Supplier No. PFLS0005BL
  • ONLINE EXCLUSIVE PRICE
    54.79€
    44.30€ / paquete de 1
    Ahorro 10.49€
    19% Descuento
Fecha estimada de envoi: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Anti-slip rubber pad kit for Fastlane 1.5 in. drop-overs
  • Anti-slip rubber pad kit for Fastlane 1.5 in. drop-overs
  • Kit includes 18 anti-slip rubber pads which help keep cable protectors in place on concrete and other hard or slick surfaces
Specifications
For Use With (Equipment) FL1X1.5 Fastlane 1.5'' Drop-Overs
Color Black
Material Polyethylene
Type Rubber Pad Kit