missing translation for 'onlineSavingsMsg'
Learn More

FANCB Antibody (M38P3E10), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-52406AF532

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215173

  • 509.71€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

FANCB Monoclonal specifically detects FANCB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

FANCB
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
FANCB
Ovalbumin-conjugated synthetic peptide- DRRENIHPYR
0.1 mL
Primary
Human
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
M38P3E10
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
EC 2.8.1, EC 3.6.3.14, FA2, FAAP90, FAAP95FAB, FACB, Fanconi anemia group B protein, Fanconi anemia, complementation group B, Fanconi anemia-associated polypeptide of 95 kDa, FLJ34064, Protein FACB
Mouse
Protein A purified
Cancer, DNA Repair, Epigenetics
2187
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales