Learn More
Novus Biologicals™ FAM216B Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-08162
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214253
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
The FAM216B Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC21784 and based on accession number NM_18258. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens chromosome 13 open reading frame 30 (C13orf30), mRNA.
Native