missing translation for 'onlineSavingsMsg'
Learn More

FAM164C Rabbit anti-Human, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 20520-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16820065

  • 319.00€ / 150 microlitros

Envío estimado: 28-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

FAM164C, also named as ZC2HC1C or C14orf140, is a 456 amino acid protein, which contains one C2HC-type zinc finger and belongs to the ZC2HC1 family. FAM164C exists as two isoforms. The isoform1 is a 51 kDa protein ad isoform2 is 31 kDa.
Especificaciones

Especificaciones

FAM164C
0.46 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q53FD0, Q6AYP4
C14orf140, FAM164C, Protein FAM164C
Rabbit
IgG
150 μL
-20°C
Primary
314321, 79696
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ZC2HC1C
Liquid
Zc2hc1c
FAM164C Fusion Protein Ag14359
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat
Videos
SDS
Documentos
Certificados