missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM125A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25816-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
FAM125A, also named as MVB12A and CFBP, is a component of the ESCRT-I complex. FAM125A is required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies. FAM125A has three isoforms with MW 27-28 kDa and 25 kDa.Especificaciones
FAM125A | |
Polyclonal | |
Unconjugated | |
MVB12A | |
CFBP, ESCRT I complex subunit MVB12A, FAM125A, Protein FAM125A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
93343 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96EY5 | |
MVB12A | |
FAM125A Fusion Protein Ag22919 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |