missing translation for 'onlineSavingsMsg'
Learn More

FABP6 Antibody (201), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90127G

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215449

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

FABP6 Monoclonal specifically detects FABP6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

FABP6
Monoclonal
DyLight 488
fatty acid binding protein 6, ileal, Fatty acid-binding protein 6, gastrotropin, I-15PIntestinal 15 kDa protein, I-BABPIntestinal bile acid-binding protein, I-BALB, I-BAP, ILBP3, ILBPGT, ileal bile acid binding protein, ILLBPIleal lipid-binding protein, illeal lipid-binding protein
This antibody was obtained from a rabbit immunized with purified, recombinant Human FABP6 (AAH22489.1; Met1-Ala128).
0.1 mL
Biologically Active Proteins, Cell Cycle and Replication
2172
Store at 4°C in the dark.
IgG
Flow Cytometry, Immunofluorescence
201
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers