missing translation for 'onlineSavingsMsg'
Learn More

FABP3/H-FABP Antibody (9F10), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-60234C

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214650

  • 562.62€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

FABP3/H-FABP Monoclonal antibody specifically detects FABP3/H-FABP in Human samples. It is validated for Western Blot, ELISA
Especificaciones

Especificaciones

FABP3/H-FABP
Monoclonal
DyLight 650
FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP
Recombinant human FABP3/H-FABP (1-133aa) purified from E. coli
0.1 mL
Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism
2170
Store at 4°C in the dark.
IgG1 κ
Western Blot, ELISA
9F10
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales