missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Antigen Rat, Biotin, Clone: BM8.1, enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB68-B-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
F4/80 Antigen | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Adgre1 | |
Protein A/G Purified | |
RUO | |
13733 | |
IgG2b κ |
Flow Cytometry | |
BM8.1 | |
Biotin | |
XP_006523664 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |