Learn More
F4/80 Antibody (CI-A3-1), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
Marca:  Novus Biologicals NB600-404G
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18216165
  • 622.01€ / 125 microlitros
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
F4/80 Monoclonal specifically detects F4/80 in Human, Mouse, C. elegans, E. coli samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
F4/80
Monoclonal
DyLight 488
ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3
Rat
160 kDa
0.125 mL
Immune System Diseases, Immunology, Innate Immunity
2015
Human, Mouse, C. elegans, E. coli
Purified
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
CI-A3-1
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
ADGRE1
Thioglycollate stimulated peritoneal macrophages from C57BL/6 mice.
Protein G purified
RUO
Primary
This recognizes mouse F4/80, a 160kD glycoprotein expressed by murine macrophages.
Store at 4C in the dark.
IgG2b