Learn More
Lenz Laborglasintrumente Extractor Heads Acc. to Thielepape
Product Code. 10274241
Change view
Click to view available options
Capacity (Metric):
150 mL
250 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10274241 250 mL Each
10040241 150 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10274241 Supplier Lenz Laborglasintrumente Supplier No. 05405149
  • ONLINE EXCLUSIVE PRICE
    341.00€
    229.37€ / paquete de 1
    Ahorro 111.63€
    32% Descuento
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Extractor heads acc. to Thielepape
  • With NS 2-way stopcock
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass 3.3
Standard Taper Joint NS 45/40