Learn More
Lenz Laborglasintrumente Extractor Heads Acc. to Soxhlet
Product Code. 10121571
Change view
Click to view available options
Capacity (Metric):
70 mL
100 mL
200 mL
250 mL
300 mL
500 mL
1 L
Standard Taper Joint:
NS 34/35
NS 45/40
NS 60/46
NS 71/51
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Standard Taper Joint Unit Size
10121571 500 mL NS 60/46 Each
10047910 200 mL NS 45/40 Each
10681702 70 mL NS 34/35 Each
10151521 100 mL NS 45/40 Each
10558421 250 mL NS 45/40 Each
10663511 300 mL NS 60/46 Each
10444451 1 L NS 71/51 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10121571 Supplier Lenz Laborglasintrumente Supplier No. 05400158
  • ONLINE EXCLUSIVE PRICE
    297.00€
    201.41€ / paquete de 1
    Ahorro 95.59€
    32% Descuento
Fecha estimada de envoi: 08-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Extractor heads acc. to DIN 12602
  • Available with/without NS stopcock
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass 3.3
Standard Taper Joint NS 60/46