Learn More
Gibco™ Expi293F™ GnTI- Cells
Product Code. 16346552
Change view
Click to view available options
Quantity:
1 vial
Unit Size:
Each
Product Code. Quantity Unit Size
16346552 1 vial Each
1 options
This item is not returnable. View return policy
Product Code. 16346552 Supplier Gibco™ Supplier No. A39240
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Expi293F GnTI- cell line is derived from engineered Expi293F cells that do not have N-acetylglucosaminyltransferase I (GnTI) activity and therefore lack complex N-glycans
Powerful tool for the high-yield expression of homogeneously glycosylated recombinant proteins.
Features:
  • Suspension, high-density culture in Expi293 Expression Medium
  • Homogeneous N-glycosylation of expressed proteins
  • Equivalent protein yields to parental Expi293F cells—up to 1 g/L
  • Significantly higher protein yields than other HEK293 GnTI- cell line alternatives
Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Expi293 Expression Medium and 10% DMSO. The cells should be thawed directly into Expi293 Expression Medium
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 1 vial Expi293F GnTI- Cells (1x107 cells/vial), store in vapor-phase liquid nitrogen
Product Line Expi293
Quantity 1 vial
Shipping Condition Dry Ice
For Research Use Only. Not for use in diagnostic procedures.