Description
- Excellence Refractometer
- Offers fast refractive index measurements in the range with automatic temperature control
- Simple One Click™ Operation enables users to start a complete workflow with one click including sampling, measuring, rinsing, drying, and SOPs.
- The user management system allows for different level of access to be defined, with each user having their own personalized home screen and language.
- Measurement limits can be set for color-coded pass/fail information for accepted and rejected samples.
- Can be upgraded to a dedicated automatic multi-parameter system combining density, refractive index, pH, color, titration and more to save time, increase data quality and prevent any alteration of samples between individual analyses.
Specifications
Specifications
| Type | Refractometer |
| Scale Range | 1.32 to 1.70 nD |
| Refractive Index | 1.32 to 1.70 |
| Description | R4 offers fast refractive index measurements |
| Sample Type | Liquids |
| Accuracy | ±0.0001 nD |
| Resolution | 0.0001 |
| Display | 7 in. display with color touchscreen |
| Scale Type | nD, Brix, Inverted Sugar, Salt, Zeiss |
| For Use With (Application) | Measure Brix or Refractive Index of a sample |
| Show More |