Learn More
EVOS™ Onstage Vessel Holder, one 100-mm Petri dish
EVOS™ Onstage Vessel Holder, one 100-mm Petri dish
Marca:  Invitrogen AMEPVH037
Detalles adicionales : Peso : 2.00000kg
Código de producto. 15309661
  • 326.00€ / cada uno
Fecha de envío estimada
del proveedor 05-11-2019
Añadir a la cesta
Descripción
Descripción
EVOS™ Onstage vessel holders maximize your choice of cell culture vessels when conducting time-lapse imaging experiments using the EVOS™ FL Auto Imaging System equipped with an EVOS™ Onstage Incubator. EVOS™ Onstage vessel holders allow a perfect fit of your micro-well plates or cell culture flasks/dishes to the master plate of the EVOS™ Onstage Incubator for increased precision in sample alignment. This particular vessel holder accommodates one 100-mm Petri dish. With a snug and secure fit, yet readily switched, these vessel holders make time-lapse imaging convenient and easy.
Learn more about the EVOS™ FL Auto Imaging System.
Explore the entire EVOS™ line of imaging systems and accessories.
Especificaciones
Especificaciones
EVOS™ FL auto imaging system, EVOS™ Onstage incubator
Onstage Vessel Holder, Allow a Perfect Fit of Micro-well Plates or Cell Culture Flasks/Dishes to the Master Plate of the EVOS™ Onstage Incubator for Increased Precision in Sample Alignment
1 Each
Onstage Vessel Holder
Videos
Documentos
For Research Use Only. Not for use in diagnostic procedures.