Learn More
EVOS™ EVOS™ Vessel Holder for Automated Stage, two 25 x 75 mm slides
For use with automated stage of the EVOS™ FL Auto Imaging System
Marca:  EVOS™ AMEPVH021
Detalles adicionales : Peso : 1.00000kg
Código de producto. 15230211
  • 361.00€ / cada uno
Fecha de envío estimada
del proveedor 23-06-2020
Añadir a la cesta
Descripción
Descripción
  • Allows user to utilize different types of vessels for imaging
  • Fits microscope slide, cell culture flask or dish, or micro-well plate to stage of EVOS imaging systems for increased precision in sample alignment
  • Includes easy-to-change stage plates and vessel holders for this instrument platform
Especificaciones
Especificaciones
EVOS™
1 holder
EVOS® XL Core Imaging System, EVOS® FL Color Imaging System, EVOS® XL Imaging System, EVOS® FL Imaging System, EVOS® FL Auto Imaging System
2 25 x 75 mm microscope slides