Learn More
Thermo Scientific™ Eutech™ Sensor for TDSTestr11
Product Code. 10569672
Change view
Click to view available options
For Use With (Equipment):
TDSTestr11
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
10569672 TDSTestr11 Each
1 options
This item is not returnable. View return policy
Product Code. 10569672 Supplier Thermo Scientific™ Supplier No. 01X229713
  • ONLINE EXCLUSIVE PRICE
    93.60€
    88.30€ / paquete de 1
    Ahorro 5.30€
    5% Descuento
Fecha estimada de envoi: 15-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Get quick and accurate Total Dissolved Solids (TDS) measurements in the field, laboratories, and schools.
Eutech™ offers meters that allow the direct reading of TDS values. Eutech™ Sensor for TDSTestr11 is a replacement sensor for TDSTestr11 series, dip type. Get quick and accurate Total Dissolved Solids (TDS) measurements in the field, laboratories, and schools.
General applications:
  • water and wastewater treatment
  • boiler blow-down
  • electroplating rinse tanks
  • drinking water
  • hydroponics
  • printing industry
  • aquaculture, aquariums, fish farms
  • swimming pools and spas
TRUSTED_SUSTAINABILITY
Specifications
Type Sensor
For Use With (Equipment) TDSTestr11