Learn More
IKA™ EUROSTAR 200 Digital Overhead Stirrer
Extremely powerful laboratory stirrer for highly viscous applications and intensive mixing for quantities up to 100 L (H2O).
Marca:  IKA™ 0003990003
Detalles adicionales : Peso : 4.60000kg
 Ver más versiones de este producto
Código de producto. 16208171
  • 2568.00€ / cada uno
Fecha estimada de envío
02-12-2020
Añadir a la cesta
Descripción
Descripción
  • Automatically adjusts the speed through microprocessor controlled technology within the speed range
  • Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions
  • Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically
  • Guarantees a constant speed even with changes in viscosities of the sample
  • Digital speed display
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection
  • Short-term overload operation
  • Slim casing
  • Quiet operation
  • Error code display
Especificaciones
Especificaciones
7 Segment LED
5 to 40°C
6 to 2000 rpm
130 W
100 L
CH Plug
50/60 Hz
EUROSTAR 200 Digital
100 to 115 V, 100 to 230 V
4.6 kg
209 x 91 x 274 mm