Learn More
Eppendorf™ Research™ plus EU-IVD Single Channel Variable Volume Pipette 3-pack
The ultra light Eppendorf Research plus pipette meets the highest needs in precision and accuracy – combined with ultimate ergonomics and increased flexibility.
Marca:  Eppendorf™ 3120000925
Detalles adicionales : Peso : 2.12600kg
 Ver más versiones de este producto
Código de producto. 10408474
  • 651.00€ / 1 set
Fecha estimada de envío
06-02-2020
Añadir a la cesta
Descripción
Descripción
  • Air-displacement pipette for accurate pipetting of aqueous solutions
  • Feel the difference in weight and pipetting forces: the ultra light mechanical pipette is designed according to the strict criteria of the Eppendorf PhysioCare Concept and limits the strain on your hand and arm
  • Spring loaded tip cone (not available for 5 mL and 10 mL pipettes) for minimal tip attachment forces helps to reduce stress
  • Low tip ejection force of Eppendorf Research plus can be as low as 3.6 N
  • Adjust your pipette in seconds for better accuracy when pipetting various difficult liquids like ethanol or even when pipetting at high altitudes. Return to factory adjustment without calibration
  • Autoclave the entire pipette or only the lower part according to your needs to ensure contamination-free operation
Especificaciones
Especificaciones
Ergonómico/a
0.1 to 1, 0.5 to 5, 1 to 10 mL
In vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998
Caja completa de puntas de pipeta para los tres volúmenes