missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene glycol-bis-(β-aminoethylether)-N,N,N',N'-tetraacetic acid, 99.6%, MP Biomedicals™
Click to view available options
Cantidad:
10 g
25 g
500 g
Especificaciones
Nombre del producto químico o material | Ethylene glycol-bis-(β-aminoethylether)-N,N,N',N'-tetraacetic acid |
Color | Blanco |
Cantidad | 10 g |
Formula Weight (peso de la fórmula) | 380.4 |
Porcentaje de pureza | 0.996 |
Forma física | Polvo |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'