Learn More
Ethyl S-(+)-4-Cyano-3-hydroxybutyrate, TRC
Product Code. 30376175
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity Unit Size
30376175 25 g 25g
1 options
This item is not returnable. View return policy
Product Code. 30376175 Supplier Toronto Research Chemicals Supplier No. TRCE90792025G
 Restricted Item
Fecha estimada de envoi: 22-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl S-(+)-4-Cyano-3-hydroxybutyrate
CAS 312745-91-8
Molecular Formula C7 H11 N O3
Quantity 25 g
InChI Formula InChI=1S/C7H11NO3/c1-2-11-7(10)5-6(9)3-4-8/h6,9H,2-3,5H2,1H3/t6-/m0/s1
SMILES CCOC(=O)C[C@@H](O)CC#N
IUPAC Name ethyl (3S)-4-cyano-3-hydroxybutanoate
Molecular Weight (g/mol) 157.17
Formula Weight 157.0739