Learn More
Miristato de etilo, 98+ %, Thermo Scientific Chemicals
Catalog Number p-3586429
Click to view available options
Cantidad:
25 g
100 g
500 g
Envase:
Frasco de vidrio
124-06-1
C16H32O2
256.43
MFCD00008984
MMKRHZKQPFCLLS-UHFFFAOYSA-N
ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
31283
CHEBI:84849
tetradecanoato de etilo
CCCCCCCCCCCCCC(=O)OCC
124-06-1
C16H32O2
256.43
MFCD00008984
MMKRHZKQPFCLLS-UHFFFAOYSA-N
ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
31283
CHEBI:84849
tetradecanoato de etilo
CCCCCCCCCCCCCC(=O)OCC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Ethyl myristate
Punto de fusión 11.0°C to 12.0°C
Color Incolora
Densidad 0.8610g/mL
Punto de ebullición 295.0°C
Temperatura de inflamación >110°C
Espectro infrarrojo Auténtico
Intervalo de porcentaje de ensayo 98% min. (GC)
Envase Frasco de vidrio
Fórmula lineal CH3(CH2)12COOCH2CH3
Cantidad 25 g
Índice Merck 15, 6419
Gravedad específica 0.861
Información de solubilidad Solubility in water: insoluble
Formula Weight (peso de la fórmula) 256.43
Porcentaje de pureza 98+%
Forma física Líquido, se solidifica en el frío