missing translation for 'onlineSavingsMsg'
Learn More

Ethyl dibromofluoroacetate, 97%

Product Code. 11309205
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309205 25 g 25g
11399195 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309205 Supplier Thermo Scientific Alfa Aesar Supplier No. L16491.14
 Restricted Item
Fecha estimada de envoi: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 565-53-7 | C4H5Br2FO2 | 263.89 g/mol

Ethyl dibromofluoroacetate is a reagent used in the preparation of fluorinated molecules in organic synthesis. It is involved in the Reformatsky-type reaction to prepare syn-alfa-bromo-alfa-fluoro-beta-lactams by using diethylzinc. It is also used to prepare 2-fluoroaziridine-2-carboxylates by reaction with imines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl dibromofluoroacetate is a reagent used in the preparation of fluorinated molecules in organic synthesis. It is involved in the Reformatsky-type reaction to prepare syn-alfa-bromo-alfa-fluoro-beta-lactams by using diethylzinc. It is also used to prepare 2-fluoroaziridine-2-carboxylates by reaction with imines.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-53-7
Molecular Formula C4H5Br2FO2
Molecular Weight (g/mol) 263.89
MDL Number MFCD00042068
InChI Key COYRDMUQLSIAEX-UHFFFAOYSA-N
Synonym ethyl dibromofluoroacetate, dibromofluoroacetic acid ethyl ester, ethyldibromofluoroacetate, 2,2-dibromo-1-ethoxy-2-fluoroethan-1-one, acmc-1aqw2, ethyl 2,2-dibromo-2-fluoracetate, ethyl 2,2-dibromo-2-fluoro-acetate
PubChem CID 2737168
IUPAC Name ethyl 2,2-dibromo-2-fluoroacetate
SMILES CCOC(=O)C(F)(Br)Br

Specifications

Density 1.894
Boiling Point 82°C (33 mmHg)
Flash Point >110°C (230°F)
Linear Formula Br2CFCO2CH2CH3
Refractive Index 1.4635
Quantity 25 g
UN Number UN2922
Beilstein 1762048
Solubility Information Miscible with most organic solvents. Immiscible with water.
Formula Weight 263.89
Percent Purity 97%
Chemical Name or Material Ethyl dibromofluoroacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.