missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrocinamato de etilo, 99 %, Thermo Scientific Chemicals

Catalog Number p-7021030
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
953-26-4
C11H11NO4
221.212
MFCD00007380
PFBQVGXIMLXCQB-VMPITWQZSA-N
ethyl 4-nitrocinnamate, ethyl p-nitrocinnamate, ethyl 3-4-nitrophenyl acrylate, 2-propenoic acid, 3-4-nitrophenyl-, ethyl ester, cinnamic acid, p-nitro-, ethyl ester, ethyl-4-nitrocinnamate, e-ethyl 3-4-nitrophenyl acrylate, propenoic acid, 3-4-nitrophenyl-, ethyl ester, 4-nitrozimtsaeure-ethylester, ethyl 2e-3-4-nitrophenyl prop-2-enoate
1268038
(E)-3-(4-nitrofenil)prop-2-enoato de etilo
CCOC(=O)C=CC1=CC=C(C=C1)[N+](=O)[O-]
Este artículo no se puede devolver. Vea la política de devoluciones
953-26-4
C11H11NO4
221.212
MFCD00007380
PFBQVGXIMLXCQB-VMPITWQZSA-N
ethyl 4-nitrocinnamate, ethyl p-nitrocinnamate, ethyl 3-4-nitrophenyl acrylate, 2-propenoic acid, 3-4-nitrophenyl-, ethyl ester, cinnamic acid, p-nitro-, ethyl ester, ethyl-4-nitrocinnamate, e-ethyl 3-4-nitrophenyl acrylate, propenoic acid, 3-4-nitrophenyl-, ethyl ester, 4-nitrozimtsaeure-ethylester, ethyl 2e-3-4-nitrophenyl prop-2-enoate
1268038
(E)-3-(4-nitrofenil)prop-2-enoato de etilo
CCOC(=O)C=CC1=CC=C(C=C1)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Ethyl 4-nitrocinnamate
Punto de fusión 138°C to 140°C
Cantidad 5 g
Beilstein 2052533
Formula Weight (peso de la fórmula) 221.21
Porcentaje de pureza 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'