Learn More
Ethyl 4,6-dichloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals
Product Code. 15494896
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494896 250 mg 250mg
15445478 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Codice prodotto. 15494896 Fornitore Thermo Scientific Alfa Aesar N. del fornitore H59231.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
CAS: 21168-41-2 | Molecular Formula: C12H9Cl2NO2 | Molecular Weight: 270.11 g/mol
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 21168-41-2
Molecular Formula C12H9Cl2NO2
Molecular Weight (g/mol) 270.11
MDL Number MFCD00173348
InChI Key BSSNTDZRVNQGDF-UHFFFAOYSA-N
Synonym ethyl 4,6-dichloro-3-quinolinecarboxylate, 4,6-dichloroquinoline-3-carboxylic acid ethyl ester, ethyl-4,6-dichloroquinoline-3-carboxylate, 3-quinolinecarboxylic acid, 4,6-dichloro-, ethyl ester, ethyl-4, 6-dichloroquinoline-3-carboxylate, 4,6-dichloro-3-quinolinecarboxylic acid ethyl ester, 3-quinolinecarboxylicacid, 4,6-dichloro-, ethyl ester, 4,6-dichloroquinoline-3-carboxylic acid, ethyl ester
PubChem CID 1479087
IUPAC Name ethyl 4,6-dichloroquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C=CC(Cl)=CC2=C1Cl
Specifica
Quantity 250 mg
Formula Weight 270.11
Percent Purity 97%
Chemical Name or Material Ethyl 4,6-dichloroquinoline-3-carboxylate
RUO – Research Use Only
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.