missing translation for 'onlineSavingsMsg'
Learn More

4,6,8-Tricloroquinolina-3-carboxilato de etilo, 97 %, Thermo Scientific™

Catalog Number p-7039883
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
31602-08-1
C12H8Cl3NO2
304.55
MFCD00173373
HAPRHXIRCWGCIG-UHFFFAOYSA-N
3-quinolinecarboxylic acid, 4,6,8-trichloro-, ethyl ester, acmc-209hnw, ethyl 4,6,8-trichloro-3-quinolinecarboxylate
709528
4,6,8-tricloroquinolina-3-carboxilato de etilo
CCOC(=O)C1=CN=C2C(Cl)=CC(Cl)=CC2=C1Cl
Este artículo no se puede devolver. Vea la política de devoluciones
31602-08-1
C12H8Cl3NO2
304.55
MFCD00173373
HAPRHXIRCWGCIG-UHFFFAOYSA-N
3-quinolinecarboxylic acid, 4,6,8-trichloro-, ethyl ester, acmc-209hnw, ethyl 4,6,8-trichloro-3-quinolinecarboxylate
709528
4,6,8-tricloroquinolina-3-carboxilato de etilo
CCOC(=O)C1=CN=C2C(Cl)=CC(Cl)=CC2=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Ethyl 4,6,8-trichloroquinoline-3-carboxylate
Cantidad 250 mg
Formula Weight (peso de la fórmula) 304.56
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'