missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzo[b]tiofeno-2-carboxilato de etilo, 97 %, Thermo Scientific™

Catalog Number p-7032548
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
34761-09-6
C11H11NO2S
221.27
MFCD00661903
XZSFZYVMRZLUOJ-UHFFFAOYSA-N
ethyl 3-aminobenzo b thiophene-2-carboxylate, ethyl 3-aminobenzothiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 3-amino-, ethyl ester, 3-amino-benzo b thiophene-2-carboxylic acid ethyl ester, 3-aminobenzo b thiophene-2-carboxylic acid ethyl ester, maybridge1_003360, 3-amino-2-carboethoxy-benzothiophene, benzo b thiophene-2-carboxylicacid, 3-amino-, ethyl ester
731510
ethyl 3-amino-1-benzothiophene-2-carboxylate
CCOC(=O)C1=C(N)C2=CC=CC=C2S1
Este artículo no se puede devolver. Vea la política de devoluciones
34761-09-6
C11H11NO2S
221.27
MFCD00661903
XZSFZYVMRZLUOJ-UHFFFAOYSA-N
ethyl 3-aminobenzo b thiophene-2-carboxylate, ethyl 3-aminobenzothiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 3-amino-, ethyl ester, 3-amino-benzo b thiophene-2-carboxylic acid ethyl ester, 3-aminobenzo b thiophene-2-carboxylic acid ethyl ester, maybridge1_003360, 3-amino-2-carboethoxy-benzothiophene, benzo b thiophene-2-carboxylicacid, 3-amino-, ethyl ester
731510
ethyl 3-amino-1-benzothiophene-2-carboxylate
CCOC(=O)C1=C(N)C2=CC=CC=C2S1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Ethyl 3-aminobenzo[b]thiophene-2-carboxylate
Punto de fusión ∼82°C (sublimation)
Cantidad 1 g
Formula Weight (peso de la fórmula) 221.28
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'