Learn More
Ethyl 3-(2-thienyl)-1H-pyrazole-5-carboxylate, 97%, Maybridge
Marca: Maybridge SPB04031DE
Detalles adicionales : CAS : 121195-03-7 Peso : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Cantidad. | 5g |
---|---|
Envase | Amber Glass Bottle |
Identificadores de productos químicos
121195-03-7 | |
222.262 | |
NTCRDZSJYGZRIE-UHFFFAOYSA-N | |
2736478 | |
CCOC(=O)C1=NNC(=C1)C2=CC=CS2 |
C10H10N2O2S | |
MFCD07772880 | |
ethyl 5-thien-2-yl-1h-pyrazole-3-carboxylate, ethyl 3-thiophen-2-yl-1h-pyrazole-5-carboxylate, ethyl 5-2-thienyl pyrazole-3-carboxylate, ethyl 5-thiophen-2-yl-1h-pyrazole-3-carboxylate, ethyl 3-2-thienyl-1h-pyrazole-5-carboxylate, ethyl 5-thiophen-2-yl-2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, 5-2-thienyl-, ethyl ester, 1h-pyrazole-3-carboxylicacid, 5-2-thienyl-, ethyl ester, acmc-20mpcx, maybridge1_008811 | |
ethyl 5-thiophen-2-yl-1H-pyrazole-3-carboxylate |
Especificación
Ethyl 5-thien-2-yl-1H-pyrazole-3-carboxylate | |
5g | |
97.0 | |
C10H10N2O2S | |
ethyl 5-thien-2-yl-1h-pyrazole-3-carboxylate, ethyl 3-thiophen-2-yl-1h-pyrazole-5-carboxylate, ethyl 5-2-thienyl pyrazole-3-carboxylate, ethyl 5-thiophen-2-yl-1h-pyrazole-3-carboxylate, ethyl 3-2-thienyl-1h-pyrazole-5-carboxylate, ethyl 5-thiophen-2-yl-2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, 5-2-thienyl-, ethyl ester, 1h-pyrazole-3-carboxylicacid, 5-2-thienyl-, ethyl ester, acmc-20mpcx, maybridge1_008811 | |
CCOC(=O)C1=NNC(=C1)C2=CC=CS2 | |
222.262 | |
222.27 |
Amber Glass Bottle | |
121195-03-7 | |
95% | |
MFCD07772880 | |
NTCRDZSJYGZRIE-UHFFFAOYSA-N | |
ethyl 5-thiophen-2-yl-1H-pyrazole-3-carboxylate | |
2736478 | |
95% |