Learn More
Ethyl [3-(10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ylidene)propyl]methylcarbamate, Mikromol™
Product Code. 30050312
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30050312 50 mg 50mg
1 options
This item is not returnable. View return policy
Product Code. 30050312 Supplier Mikromol Supplier No. MM0079.12
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl [3-(10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ylidene)propyl]methylcarbamate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl [3-(10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ylidene)propyl]methylcarbamate
CAS 16234-88-1
Analyte or Component Names Ethyl [3-(10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ylidene)propyl]methylcarbamate
Shipping Condition Ice pack (-20°C)
Molecular Formula C22 H25 N O2
Certifications/Compliance ISO 9001
Synonym Nortriptyline Hydrochloride Imp. G (EP), Nortriptyline Imp. G (EP), Ethyl [3-(10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ylidene)propyl]methylcarbamate, Nortriptyline N-Ethyl Carbamate, Nortriptyline Hydrochloride Impurity G, Nortriptyline Impurity G
InChI Formula InChI=1S/C22H25NO2/c1-3-25-22(24)23(2)16-8-13-21-19-11-6-4-9-17(19)14-15-18-10-5-7-12-20(18)21/h4-7,9-13H,3,8,14-16H2,1-2H3
SMILES CCOC(=O)N(C)CCC=C1c2ccccc2CCc3ccccc13
IUPAC Name ethyl N-[3-(5,6-dihydrodibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)propyl]-N-methylcarbamate
Molecular Weight (g/mol) 335.44
Quantity 50 mg
Formula Weight 335.1885 g/mol